To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Solute Carrier Family 35 Member D2
- Antigen Symbol:SLC35D2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:11046
- Antigen Synonyms:UGTrel8|hfrc|HFRC1|SQV7L|HFRC|Homolog of Fringe connection protein 1|SLC35D2|UGTREL8
- Storage Buffer:PBS (0.02% sodium azide)
- Storage Temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 14 amino acids at C-terminus of human SLC35D2.
- Purification:Peptide affinity purification
- Cat. No.:10738-566
- Supplier no.:PAB25738
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of SLC35D2.
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunocytochemistry: 5 ?g/ml
Type: Primary
Antigen: SLC35D2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human