To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:NEL-like 2
- Antigen Symbol:NELL2
- Clonality:Monoclonal
- Clone:AT13E7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:4753
- Antigen Synonyms:NEL-like protein 2|Mel91|NELL2|Protein kinase C-binding protein NELL2|NRP2|MEL91 protein|A330108N19Rik|Nel-related protein 2|NELL2_HUMAN|R75516.|NEL like 2 chicken
- Amino Acid Number:30 to 258
- Storage Buffer:PBS, pH 7.4 (0.1% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 30-258 of human NELL2.
- Purification:Protein G purification
- Cat. No.:10726-926
- Supplier no.:MAB10014
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant NELL2.
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: NELL2
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: AT13E7
Epitope:
Host: IgG2b kappa
Isotype: Mouse
Reactivity: Human, Mouse