Order Entry
United States
ContactUsLinkComponent
Anti-MAPK14 Mouse Monoclonal Antibody [clone: M139]
Anti-MAPK14 Mouse Monoclonal Antibody [clone: M139]
Catalog # 10674-906
Supplier:  Abnova
undefined
Anti-MAPK14 Mouse Monoclonal Antibody [clone: M139]
Catalog # 10674-906
Supplier:  Abnova
Supplier Number:  MAB1385
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Mitogen-Activated Protein Kinase 14
  • Antigen Symbol:
    MAPK14
  • Clonality:
    Monoclonal
  • Clone:
    M139
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    81649
  • Antigen Synonyms:
    p38ALPHA|Mxi2|CSPB1|PRKM15|CSBP|CSBP2|EXIP|p38|RK|CSBP1|PRKM14|SAPK2A
  • Modification:
    phospho-Thr180/Tyr-182
  • Storage Buffer:
    PBS (50% glycerol, 1 mg/mL BSA, 0.05% sodium azide)
  • Storage Temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding T180/Y182 of rat Mapk14.
  • Cat. No.:
    10674-906
  • Supplier no.:
    MAB1385

Specifications

About this item

Mouse monoclonal antibody raised against synthetic phosphopeptide of Mapk14.

Recommended Dilutions: ELISA: 1:2000; Immunocytochemistry: 1:250; Western Blot: 1:1000

Type: Primary
Antigen: MAPK14
Clonality: Monoclonal
Clone: M139
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat