To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Mitogen-Activated Protein Kinase Associated Protein 1
- Antigen Symbol:MAPKAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:79109
- Antigen Synonyms:TORC2 subunit MAPKAP1|MAPKAP1|SIN1|MIP1|Mitogen-activated protein kinase 2-associated protein 1|JC310|SIN1b|Target of rapamycin complex 2 subunit MAPKAP1|SIN1g
- Amino Acid Number:100 to 150
- Storage Buffer:PBS (0.2% gelatin, 0.05% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 100-150 of human MAPKAP1.
- Cat. No.:10724-902
- Supplier no.:PAB0307
Rabbit polyclonal antibody raised against partial recombinant MAPKAP1.
Type: Primary
Antigen: MAPKAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Human, Mouse, Rat