To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Autophagy Related 4C
- Antigen Symbol:ATG4C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:84938
- Antigen Synonyms:APG4-C|Autophagy-related protein 4 homolog C,|AUTL3|ATG4 autophagy related 4 homolog C (S. cerevisiae)|autophagin-3|AUTL1|APG4C|ATG4C|FLJ14867
- Storage Buffer:PBS
- Storage Temperature:Store at 4 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with Blue Carrier Protein) corresponding to C-terminus of human ATG4C.
- Cat. No.:10690-390
- Supplier no.:PAB0737
Rabbit polyclonal antibody raised against synthetic peptide of ATG4C.
Recommended Dilutions: Immunohistochemistry: 1:100-1:2000; Immunofluorescence: 1:100-1:2000; Western Blot: 1:100-1:2000
Type: Primary
Antigen: ATG4C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat