To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:SPANX Family Member C
- Antigen Symbol:SPANXC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Gorilla,Chimpanzee,Human
- Western Blot:Yes
- Size:500 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Lyophilized
- Gene ID:64663
- Antigen Synonyms:C|CT11.3|CTp11|SPANX-C
- Storage Buffer:20 mM KH2PO4, 150 mM NaCl, pH 7.2 (0.01% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot after reconstitution to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human SPANXC.
- Cat. No.:10735-652
- Supplier no.:PAB11268
Rabbit polyclonal antibody raised against full length recombinant SPANXC.
Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:1000-1:5000
Type: Primary
Antigen: SPANXC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Gorilla, Human