To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:DYKDDDDK tag
- Antigen Symbol:DYKDDDDK tag
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:PBS, pH 7.2 (10% Proclin300)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to DYKDDDDK tag.
- Cat. No.:10713-356
- Supplier no.:PAB12767
Rabbit polyclonal antibody raised against synthetic peptides of DYKDDDDK tag.
Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml
Type: Primary
Antigen: DYKDDDDK tag
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: