To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Kinesin (heavy chain)
- Antigen Symbol:Kinesin (heavy chain)
- Clonality:Monoclonal
- Clone:KN-03
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgM
- Reactivity:Pig,Human,Rat,Mouse
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Storage Buffer:PBS, pH 7.4 (15 mM sodium azide)
- Storage Temperature:Store at 4°C. Do not freeze.
- Immunogen:Native purified porcine Kinesin (heavy chain).
- Cat. No.:10722-472
- Supplier no.:MAB3622
Mouse monoclonal antibody raised against native Kinesin (heavy chain).
Type: Primary
Antigen: Kinesin (heavy chain)
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: KN-03
Epitope:
Host: IgM
Isotype: Mouse
Reactivity: Human, Mouse, Pig, Rat