To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Mitogen-Activated Protein Kinase 4
- Antigen Symbol:MAPK4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross Adsorption:No
- Format:Liquid
- Gene ID:5596
- Antigen Synonyms:ERK-4|ERK4|PRKM4|p63-MAPK|p63MAPK
- Amino Acid Number:544 to 574
- Storage Buffer:PBS (0.09% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to amino acids 544-574 at C-terminus of human MAPK4.
- Purification:Ammonium sulfate precipitation
- Cat. No.:10679-240
- Supplier no.:PAB2872
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MAPK4.
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:50-100
Type: Primary
Antigen: MAPK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human