To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Protein Disulfide Isomerase Family A, Member 3
- Antigen Symbol:PDIA3
- Clonality:Monoclonal
- Clone:MaP.ERP57
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Hamster,Pig,Rabbit,Monkey,Human,Bovine,Guinea pig,Dog,Rat,Mouse
- Western Blot:Yes
- Size:200 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:2923
- Antigen Synonyms:HEL-S-269|PI-PLC|HEL-S-93n|ER60|HsT17083|ERp60|GRP57|GRP58|ERp61|ERp57|P58
- Storage Buffer:PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human PDIA3.
- Cat. No.:10676-990
- Supplier no.:MAB6645
Mouse monoclonal antibody raised against recombinant PDIA3.
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: PDIA3
Clonality: Monoclonal
Clone: MaP.ERP57
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Guinea Pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat