Order Entry
United States
ContactUsLinkComponent
 
Anti-MAP2K5 Rabbit Polyclonal Antibody
  10679-198
 :  Abnova
undefined
Anti-MAP2K5 Rabbit Polyclonal Antibody
  10679-198
 :  Abnova
 :  PAB2270
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Mitogen-Activated Protein Kinase Kinase 5
  • Antigen Symbol:
    MAP2K5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Environmentally Preferable:
  • Cross Adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    5607
  • Antigen Synonyms:
    PRKMK5|MEK5|MAPKK5|HsT17454
  • Storage Buffer:
    PBS (0.09% sodium azide)
  • Storage Temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide (conjugated with KLH) corresponding to residues surrounding S149 of human MAP2K5.
  • Purification:
    Protein A purification
  • Cat. No.:
    10679-198
  • Supplier no.:
    PAB2270

 

 

Rabbit polyclonal antibody raised against synthetic peptide of MAP2K5.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:50-100; Immunohistochemistry: 1:10-50

Type: Primary
Antigen: MAP2K5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human