To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:energy homeostasis associated
- Antigen Symbol:ENHO
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:375704
- Antigen Synonyms:C9orf165|UNQ470
- Amino Acid Number:10 to 60
- Storage Buffer:PBS (30% glycerol, 0.1% sodium azide)
- Storage Temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 10-60 of human ENHO.
- Cat. No.:10712-024
- Supplier no.:PAB15531
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of ENHO.
Recommended Dilutions: Western Blot: 2 ?g/ml
Type: Primary
Antigen: ENHO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rat