To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Carnitine Palmitoyltransferase 1B
- Antigen Symbol:CPT1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:1375
- Antigen Synonyms:CPT1B|FLJ58750|carnitine palmitoyltransferase 1B (muscle)|carnitine palmitoyltransferase 1B (muscle) |KIAA1670|CPTI|FLJ55729|carnitine palmitoyltransferase 1A (muscle)|carnitine O-palmitoyltransferase I|carnitine palmitoyltransferase I|M-CPT1|muscle|mitochondrial muscle isoform|carnitine palmitoyltransferase I-like|CPT1-M|carnitine palmitoyltransferase 1B|MCCPT1|CPT1M|MCPT1
- Storage Buffer:PBS (0.09% sodium azide)
- Storage Temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CPT1B.
- Purification:Protein G purification
- Cat. No.:10678-472
- Supplier no.:PAB4375
Rabbit polyclonal antibody raised against synthetic peptide of CPT1B.
Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:50-200; Immunohistochemistry: 1:50-100
Type: Primary
Antigen: CPT1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: