To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Aquaporin 12A
- Antigen Symbol:AQP12A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:375318
- Antigen Synonyms:AQP-12|AQPX2|AQP12
- Storage Buffer:PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
- Storage Temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal of human AQP12A.
- Cat. No.:10689-870
- Supplier no.:PAB17612
Rabbit polyclonal antibody raised against synthetic peptide of AQP12A.
Recommended Dilutions: Western Blot: 1:500-1:1000; ELISA: 1:40000
Type: Primary
Antigen: AQP12A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human