To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:G Protein-coupled Receptor 149
- Antigen Symbol:GPR149
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Pig,Monkey,Gorilla,Human
- Size:50 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Format:Liquid
- Gene ID:344758
- Antigen Synonyms:IEDA|PGR10
- Storage Buffer:PBS (0.1% sodium azide)
- Storage Temperature:Store at 4°C. For long term storage store at –80 °C . Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 15 amino acids at N-terminal extracellular domain of human GPR149.
- Purification:Immunoaffinity chromatography
- Cat. No.:10684-316
- Supplier no.:PAB26368
Rabbit polyclonal antibody raised against synthetic peptide of GPR149.
Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 7-18 ?g/ml
Type: Primary
Antigen: GPR149
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Gorilla, Human, Monkey, Pig