Anti-ZIC1 Mouse Monoclonal Antibody [clone: 1D12]
Catalog # 10606-146
Supplier: Abnova
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Zic Family member 1
- Antigen Symbol:ZIC1
- Clonality:Monoclonal
- Clone:1D12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Gene ID:7545
- Antigen Synonyms:Zic1 encodes a zinc finger protein expressed in the developing or matured central nervous system in a highly restricted manner. Zic is expressed in granule cells that make synaptic contact with Purkinje cells. Zic1 is a gene critical to cerebellar pattern formation. The expression of Zic genes is first detected at gastrulation and at neurulation|becoming restricted to the dorsal neural ectoderm and the dorsal paraxial mesoderm. The Zic1 gene has been mapped to chromosome 9 in mouse. The 5' flanking region of the Zic1 gene contains a region-specific enhancer determined to be essential in in-vivo and in-vitro deletion analysis. The temporal profile of mRNA expression differs for each of the Zic gene products. The Drosophila odd-paired gene is highly homologous to the Zic gene family. Zic2 and Zic3 are highly similar genes. Zic2 is essential for the formation of the brain and Zic-3 is important for right and left axis formation.
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10606-146
- Supplier no.:H00007545-M04
Specifications
About this item
Type: Primary
Antigen: ZIC1
Clonality: Monoclonal
Clone: 1D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human