Anti-TOP2A (topoisomerase (DNA) II alpha 170kDa) Mouse Monoclonal Antibody [clone: 100]
Catalog # 10604-404
Supplier: Abnova
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Topoisomerase 2 alpha
- Antigen Symbol:TOP2A
- Clonality:Monoclonal
- Clone:1E2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Cross Adsorption:No
- Gene ID:7153
- Antigen Synonyms:alpha isozyme|DNA topoisomerase 2-alpha|DNA gyrase|DNA topoisomerase (ATP-hydrolyzing)|TP2A|170 kD|DNA topoisomerase II|TOP2
- Storage Temperature:Cool
- Shipping Temperature:Ice
- Cat. No.:10604-404
- Supplier no.:H00007153-M01
Specifications
About this item
Type: Primary
Antigen: TOP2A (topoisomerase (DNA) II alpha 170kDa)
Clonality: Monoclonal
Clone: 100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human