To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Mannosidase Alpha Class 1B Member 1
- Antigen Symbol:ERManI
- Clonality:Monoclonal
- Clone:3C2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:11253
- Antigen Synonyms:MRT15|alpha|RP11-350O14.2|ER alpha-1,2-mannosidase|Mannosidase alpha class 1B member 1|mannosidase|ERMan1|ER alpha 1,2-mannosidase|endoplasmic reticulum alpha-mannosidase 1|member 1|MANA-ER|MAN1B1|alpha 1,2-mannosidase|Man9GlcNAc2-specific processing alpha-mannosidase|EC 3.2.1.113|ER mannosidase 1|endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase|ER ManI|class 1B|endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase 1|Man9GlcNAc2-specific-processing alpha-mannosidase
- Storage Buffer:Tris-Glycine (pH 7.5) and 0.15M NaCl
- Storage Temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:1.0 mg/ml
- Shipping Temperature:Ice
- Immunogen:Full length human recombinant ERManI [Swiss-Prot# Q9UKM7]
- Purification:Protein G purified
- Cat. No.:102715-814
- Supplier no.:NBP2-13167
The ERManI Antibody (3C2) from Novus Biologicals is a mouse monoclonal antibody to ERManI. This antibody reacts with human, mouse (negative). The ERManI Antibody (3C2) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Knockout Validated.
Type: Primary
Antigen: ERManI
Clonality: Monoclonal
Clone: 3C2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse (Negative)