To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Cerebellar Degeneration-Related Protein 2, 62Kda
- Antigen Symbol:CDR2
- Clonality:Monoclonal
- Clone:33
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western Blot:Yes
- Size:100 μl
- Environmentally Preferable:
- Gene ID:1039
- Antigen Synonyms:62kDa|CDR62|Paraneoplastic cerebellar degeneration-associated antigen|Yo|cerebellar degeneration-related protein 2|PCD17|Major Yo paraneoplastic antigen|Yo paraneoplastic antigen|cerebellar degeneration-related protein (62kD)
- Storage Buffer:PBS
- Storage Temperature:Store at 4C in the dark.
- Shipping Temperature:Ice
- Immunogen:N-terminal MBP-tagged full length human CDR2 [Swiss-Prot # Q01850]
- Purification:Protein G purified
- Cat. No.:102715-738
- Supplier no.:NBP2-10509B
The CDR2 Antibody (33) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to CDR2. This antibody reacts with human, mouse (negative). The CDR2 Antibody (33) [Biotin] has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: CDR2
Clonality: Monoclonal
Clone: 33
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse (Negative)