To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:beta-1,3-Glucuronyltransferase 1/B3GAT1
- Antigen Symbol:B3GAT1
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 μl
- Environmentally Preferable:
- Gene ID:27087
- Antigen Synonyms:EC 2.4.1.135|NK1|NK-1|GlcAT-P|GLCATPGLCUATP|beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P)|Beta-1,3-glucuronyltransferase 1|Glucuronosyltransferase P|UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase|glcUAT-P|galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1
- Storage Buffer:50mM Sodium Borate
- Storage Temperature:Store at 4C in the dark.
- Shipping Temperature:Ice
- Immunogen:A synthetic peptide made to a C-terminal portion of the human B3GAT1 protein (between residues 250-300) [UniProt Q9P2W7]
- Purification:Immunogen affinity purified
- Cat. No.:102715-576
- Supplier no.:NBP1-78982G
The beta-1,3-Glucuronyltransferase 1 / B3GAT1 Antibody [DyLight 488] from Novus Biologicals is a rabbit polyclonal antibody to beta-1,3-Glucuronyltransferase 1 / B3GAT1. This antibody reacts with human, mouse. The beta-1,3-Glucuronyltransferase 1 / B3GAT1 Antibody [DyLight 488] has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: beta-1,3-Glucuronyltransferase 1/B3GAT1
Clonality: Polyclonal
Clone:
Conjugation: DyLight 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse