ContactUsLinkComponent
Epredia™ Cytospin™ 4 Cytocentrifuge, Epredia
  89404-828
undefined
Epredia™ Cytospin™ 4 Cytocentrifuge, Epredia
  89404-828
 :  89404-828
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Cytospin™ 4 Cytocentrifuge
  • Dimensions:
    62L×40.5W×9.5H cm (24.5×16×25")
  • Environmentally Preferable:
  • Cat. No.:
    89404-828
  • Electrical:
    100/240 V, 50/60 Hz
  • Weight:
    12 kg (26.5 lbs.)
  • Supplier no.:
    A78300003
Gain all the advantages of the ultimate thin–layer cell preparation system with the Epredia™ Cytospin™ 4 Cytocentrifuge. This reliable benchtop centrifuge provides economical thin–layer preparations from any liquid matrix, especially hypocellular fluids such as spinal fluid and urine.
  • Processes 12 specimens at one time
  • Accepts all protocols from Cytospin 1, 2 and 3
  • Allows for one–handed opening and closing with a redesigned lid–release mechanism
  • Enables viewing of the sealed head through the polycarbonate window during operation
  • Protects mechanical and electronic components from damage due to accidental fluid spills
  • Designed for easy disinfection
  • Meets stringent IEC 61010 safety standards
  • Allows loading of the sealed head inside a microbiological cabinet for ultimate safety
  • Features an auto–locking, plastic outer lid
  • Operates only when lid is locked and keeps lid locked during rotation
Includes disposable sample chambers with caps and a wipe–clean control panel
Cytocentrifugation constructively flattens cells for excellent nuclear presentation. Spinning action tilts the Epredia™ Cytofunnels™ upright and centrifuges cells onto the deposition area of the slide. Gives all cell types equal opportunity for presentation. Tilting feature reduces cell loss by preventing residual fluid from coming into contact with the prepared slide.
Pull–out program card logs up to nine programs for fast, convenient retrieval. Protected program memory stores up to 23 routines for instant recall and protects from power loss. Easy–touch program keypad allows for easier keystrokes and more intuitive programming. Speed window displays programmed spinning speeds from 200 to 2000rpm. Three acceleration–control buttons allow the operator the flexibility to protect fragile samples. Time window displays the programmed and remaining run time from 1 to 99. Control feature has a standby power–saving mode; displays are also turned on and off by the standby key.
Sounds an audible alarm when the cytocentrifuge is out of balance, outside the speed tolerance or if the lid is not properly locked. Uses specimen safety alarms at one–minute intervals to remind users to remove specimens to prevent air drying and ensure consistent results. Deposits cells onto a clearly defined area of a glass slide and allows for the absorption of the residual fluid into the sample chamber's filter card.
 : Cytospin 4: Seated head/rotor; Marking pen; Operator's manual; Service record book; Standard workflow diagram. Epredia™ EZ Funnel Starter Kit: 25 EZ Megafunnels, 20 EZ Single White Cytofunnels, 20 EZ Double Cytofunnels and Cytoslides. Epredia™ Clipped Funnel Starter Kit: 50 Single White Cytofunnels, 25 Double Cytofunnels, Cytoslides, 12 Cytoclips.