To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Heat Shock 10kDa Protein 1
- Antigen Symbol:CPN10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Pig,Rabbit,Xenopus,Sheep,Human,Bovine,Guinea pig,Dog,Rat,Mouse
- Western Blot:Yes
- Size:50 µg
- Environmentally Preferable:
- Epitope:C-Terminal
- Form:Liquid
- Antigen Synonyms:Chaperonin 10
- Storage Buffer:PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
- Storage Temperature:–20 °C
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Synthetic peptide corresponding to the sequence near the C-terminus of human Cpn10.
- Purification:purified by Protein A affinity chromatography
- Cat. No.:10049-484
- Supplier no.:ADI-SPA-110-D
Chaperonin 10 (Cpn10 or Hsp10), is the ~10 kDa mammalian equivalent to GroES of E. coli. Chaperonins play fundamental roles in the folding, assembly, and translocation of other proteins. Chaperonin 10 has been located in human platelets but more importantly it may also be present in human maternal serum. It has been reported that human Cpn10 is identical with Early pregnancy factor (EPF), which is involved in control over cell growth and development.
Type: Primary
Antigen: CPN10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: