To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Kallikrein 8
- Antigen Symbol:KLK8
- Clonality:Monoclonal
- Clone:CGA
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Antigen Synonyms:human kallikrein 8|mK8|ovasin|tumor-associated|hK8|PRSS19|KLK8
- Amino Acid Number:25 to 260
- Storage Buffer:1X PBS, pH 7.4, containing 10% glycerol and 0.05% sodium azide.
- Storage Temperature:–20 °C
- Shipping Temperature:Shipped on Blue Ice
- Immunogen:Recombinant protein corresponding to aa 25-260 of human kallikrein-8 (His-tagged).
- Purification:purified by Protein G affinity chromatography
- Cat. No.:10049-264
- Supplier no.:905-1002-0100
Specifications
About this item
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. KLK8 is one of the fifteen kallikrein subfamily member. The KLK8 enzyme exhibits distinct patterns of expression that suggest roles in brain plasticity and ovarian cancer.
Type: Primary
Antigen: KLK8
Clonality: Monoclonal
Clone: CGA
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human