To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Angiopoietin-like 3
- Antigen Symbol:ANGPTL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Epitope:near the N-terminus
- Cross Adsorption:No
- Form:Lyophilized
- Antigen Synonyms:ANGPTL3
- Amino Acid Number:32 to 48
- Sequence:EPKSRFAMLDDVKILAN
- Storage Temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human ANGPTL3 (32-48aa EPKSRFAMLDDVKILAN), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Cat. No.:10206-658
- Supplier no.:PA2075
Rabbit IgG polyclonal antibody for Angiopoietin-related protein 3(ANGPTL3) detection. Tested with WB in Human;Mouse;Rat.
Reconstitute with distilled water
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Type: Primary
Antigen: ANGPTL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat