ContactUsLinkComponent
IHC Enzyme Antigen Retrieval Reagent, Enzo Life Sciences
IHC Enzyme Antigen Retrieval Reagent, Enzo Life Sciences
Catalog # 10049-784
Supplier:  Enzo Life Sciences
CAS Number:  
IHC Enzyme Antigen Retrieval Reagent, Enzo Life Sciences
Catalog # 10049-784
Supplier:  Enzo Life Sciences
Supplier Number:  950-280-0015
CAS Number:  
Specifications
  • Description:
    IHC Enzyme Antigen Retrieval Reagent
  • Size:
    15 mL
  • Environmentally Preferable:
  • Cat. No.:
    10049-784
  • Supplier no.:
    950-280-0015
Specifications
About this item
IHC enzyme antigen retrieval reagent is designed to unmask immunoreactive sites altered by fixation and/or the embedding process.
Liquid. Enzyme antigen retrieval reagent.