To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:ZFP36 RING Finger protein-like 1
- Antigen Symbol:ZFP36L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:150 µL
- Environmentally Preferable:
- Antigen Synonyms:BRF1|TIS11B|RNF162B|Berg36|cMG1|ERF1|ERF-1
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:ZFP36L1
- Purification:Antigen affinity purification
- Cat. No.:10156-210
- Supplier no.:12306-1-AP
The ZFP36L1 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human ZFP36L1. This antibody recognizes human,mouse,rat antigen. The ZFP36L1 antibody has been validated for the following applications: ELISA, WB, IHC, IF analysis.
WB 1 : 500 ; IHC 1 : 50
Type: Primary
Antigen: ZFP36L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat