Order Entry
United States
ContactUsLinkComponent
GHS Reference Center Poster, Brady®
GHS Reference Center Poster, Brady®
Catalog # 10154-260
Supplier:  Brady Worldwide
undefined
GHS Reference Center Poster, Brady®
Catalog # 10154-260
Supplier:  Brady Worldwide
Supplier Number:  133209

Specifications

  • Language:
    English
  • Color:
    Brown
  • Description:
    GHS Reference Center Poster
  • Dimensions:
    39.5×25.5"
  • Material:
    Paper
  • Environmentally Preferable:
  • Cat. No.:
    10154-260
  • Supplier no.:
    133209

Specifications

About this item

This all inclusive poster is great for combining many of the most important changes into one easy-to-read format and reference poster combines new Safety Data Sheet format and sections, new GHS-formatted label elements and GHS Hazard Pictograms.

  • Easy-to-read, large format poster gets workers attention
  • Lamination protects graphics and improves durability
  • Posters are perfect for employee training and daily reminders of most important GHS standard changes