ContactUsLinkComponent
Diamond-Tipped Glass Cutter, Chemglass
Diamond-Tipped Glass Cutter, Chemglass
Catalog # 80060-586
Supplier:  Chemglass
Small Business Enterprise
undefined
Diamond-Tipped Glass Cutter, Chemglass
Catalog # 80060-586
Supplier:  Chemglass
Supplier Number:  CG-1179-20
Small Business Enterprise
Specifications
  • Description:
    Glass Cutter with Turet Head
  • Environmentally Preferable:
  • Cat. No.:
    80060-586
  • Supplier no.:
    CG-1179-20
Specifications
About this item
Nickel-plated brass plate glass cutter has a diamond tip with a red wooden handle.
The rack has four slots that will accommodate plates from 1/16" to 3/16" (1.5 to 4.75 mm) thick.