To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Bath Capacity:270 - 320 ml
- Memory:Yes
- Description:MUPID One Horizontal Electrophoresis System with Attached Power Supply
- Environmentally Preferable:
- L×W×H:162 mm x 183 mm x 59 mm
- Output voltage:18 V, 25 V, 35 V, 50 V, 70 V, 100 V and 135 V.
Constant peak voltage of 140 V and duty control - Safety:Yes
- Timer:0 and 99 min
- Cat. No.:102407-972
- Supplier no.:MU2
Specifications
About this item
The Mupid-One Electrophoresis System is a convenient DNA separation systems
It has many novel features including a separated power supply, a simple buffer drainage system, support for multi-channel pipettes ,and seven output voltage settings as well as a timer function for delivering the perfect run every time. 1 year warranty.