Gold Heat Reflective Face Shields, Oberon
Supplier: Sellstrom
Certificates
About this item
The Oberon Gold Reflective Faceshield is designed for high radiant heat applications.
- Heat reflective series of faceshields is manufactured using space-age technology
- Coated with a mirror-like surface of 24K Gold to reflect the radiant energy away from the user’s face
- Headgear is ratchet adjustable and comfortable
- Ideal for shop, industrial, and laboratory use
Ordering information: Contact your Avantor sales representative for more information.