To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:GAR1 ribonucleoprotein
- Antigen Symbol:GAR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:0.05 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9NY12
- Antigen Synonyms:GAR1
- Storage Buffer:Phosphate-Buffered Saline. No preservatives added.
- Molecular Weight:22.3 kDa (calculated)
- Storage Temperature:NOLA1 antibody can be stored at 4 °C for short term (weeks). Long term storage should be at –20 °C. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:1-217
- Tested Applications:Immunoaffinity Purified
- Purification:Immunoaffinity Purified
- Cat. No.:10069-756
- Supplier no.:XW-7880
Specifications
About this item
FUNCTION: Required for ribosome biogenesis and telomere maintenance. Part of the H/ACA small nucleolar ribonucleoprotein (H/ACA snoRNP) complex, which catalyzes pseudouridylation of rRNA. This involves the isomerization of uridine such that the ribose is subsequently attached to C5, instead of the normal N1. Each rRNA can contain up to 100 pseudouridine ("psi") residues, which may serve to stabilize the conformation of rRNAs. May also be required for correct processing or intranuclear trafficking of TERC, the RNA component of the telomerase reverse transcriptase (TERT) holoenzyme.
H/ACA ribonucleoprotein complex subunit 1 antibody can be used for the detection of H/ACA ribonucleoprotein complex subunit 1 by Western Blot.
Phosphate-Buffered Saline. No preservatives added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: GAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human, Mouse