Anti-BRWD1 Chicken Polyclonal Antibody
Catalog # 10068-108
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:bromodomain and WD repeat domain containing 1
- Antigen Symbol:BRWD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:0.5 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9NSI6
- Antigen Synonyms:BRWD1
- Storage Temperature:2-8 °C for one week, –20 °C as an undiluted liquid for up to two year.
- Shipping Temperature:4 °C
- Immunogen:WDR9.3 protein, 57 kDa
- Cat. No.:10068-108
- Supplier no.:XG-6115
Specifications
About this item
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: BRWD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype:
Reactivity: Human