Order Entry
United States
ContactUsLinkComponent
Anti-AKT1S1 Rabbit Polyclonal Antibody
Anti-AKT1S1 Rabbit Polyclonal Antibody
Catalog # 10062-560
Supplier:  Prosci
undefined
Anti-AKT1S1 Rabbit Polyclonal Antibody
Catalog # 10062-560
Supplier:  Prosci
Supplier Number:  7391
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    AKT1 substrate 1 (proline-rich)
  • Antigen Symbol:
    AKT1S1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.1 mg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    Q96B36
  • Antigen Synonyms:
    AKT1S1
  • Storage Buffer:
    AKT1S1 Antibody is supplied in PBS containing 0.02% sodium azide.
  • Molecular Weight:
    Predicted: 30 kDa Observed: 28 kDa
  • Storage Temperature:
    AKT1S1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    AKT1S1 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human AKT1S1. The immunogen is located within amino acids 190 - 240 of AKT1S1.
  • Purification:
    AKT1S1 Antibody is affinity chromatography purified via peptide column.
  • Cat. No.:
    10062-560
  • Supplier no.:
    7391

Specifications

About this item

AKT1S1 Antibody: The Akt signaling pathway contributes to the regulation of apoptosis after a variety of cell death signals. AKT1S1, also known as PRAS40, is a proline-rich substrate of the kinase AKT1 and is thought to play a role in neuroprotection mediated by nerve growth factor (NGF) after transient focal cerebral ischemia. AKT1S1 is also a substrate and potential regulator of mammalian target of rapamycin (mTOR), a serine/threonine kinase that regulates cell growth and cell cycle, and a negative regulator of autophagy. Treatment with the insulin-like growth factor-1 (IGF1) can indusce the phosphorylation of AKT1S1 via the PI3K/AKT signaling pathway in PC12 cells.

AKT1S1 antibody can be used for detection of AKT1S1 by Western blot at 1 - 2 ?g/mL.

AKT1S1 Antibody is affinity chromatography purified via peptide column.

AKT1S1 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AKT1S1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat