Order Entry
United States
ContactUsLinkComponent
Anti-DUSP14 Goat Polyclonal Antibody
Anti-DUSP14 Goat Polyclonal Antibody
Catalog # 10112-506
Supplier:  Prosci
undefined
Anti-DUSP14 Goat Polyclonal Antibody
Catalog # 10112-506
Supplier:  Prosci
Supplier Number:  45-505
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    dual specificity phosphatase 14
  • Antigen Symbol:
    DUSP14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
  • Size:
    0.1 mg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    O95147
  • Antigen Synonyms:
    DUSP14
  • Storage Buffer:
    DUSP14 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
  • Storage Temperature:
    Aliquot and store at –20 °C. Minimize freezing and thawing.
  • Concentration:
    500 ug/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    DUSP14 antibody was raised against a 14 amino acid synthetic peptide near the C-Terminus of DUSP14.
  • Purification:
    DUSP14 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Cat. No.:
    10112-506
  • Supplier no.:
    45-505

Specifications

About this item

ELISA: Antibody detection limit dilution 1:32000. Western Blot: No signal obtained yet but low background observed in human liver extracts at up to 1 ?g/mL.

DUSP14 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DUSP14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: