Order Entry
United States
ContactUsLinkComponent
Anti-ADH5 Goat Polyclonal Antibody
Anti-ADH5 Goat Polyclonal Antibody
Catalog # 10112-338
Supplier:  Prosci
undefined
Anti-ADH5 Goat Polyclonal Antibody
Catalog # 10112-338
Supplier:  Prosci
Supplier Number:  45-207
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    alcohol dehydrogenase 5 (class III), chi polypeptide
  • Antigen Symbol:
    ADH5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.1 mg
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    P11766
  • Antigen Synonyms:
    ADH5
  • Storage Buffer:
    ADH5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
  • Storage Temperature:
    Aliquot and store at –20 °C. Minimize freezing and thawing.
  • Concentration:
    500 ug/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    ADH5 antibody was raised against a 12 amino acid synthetic peptide near the internal region of ADH5.
  • Purification:
    ADH5 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Cat. No.:
    10112-338
  • Supplier no.:
    45-207

Specifications

About this item

ELISA: Antibody detection limit dilution 1:1,000. Western Blot: Customer's experiments gave a band at approximately 38 kDa in mouse Lymphocyte lysates (calculated MW of 39.7 kDa according to human NP_000662.3 and 39.5 kDa according to mouse NP_031436.2). Recommended concentration: 1-3 ?g/mL.

ADH5 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ADH5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat