Order Entry
United States
ContactUsLinkComponent
Anti-TYRO3 Mouse Monoclonal Antibody [clone: 1C10E8 / 9A3F2]
Anti-TYRO3 Mouse Monoclonal Antibody [clone: 1C10E8 / 9A3F2]
Catalog # 10070-706
Supplier:  Prosci
undefined
Anti-TYRO3 Mouse Monoclonal Antibody [clone: 1C10E8 / 9A3F2]
Catalog # 10070-706
Supplier:  Prosci
Supplier Number:  32-233
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    TYRO3 protein tyrosine kinase
  • Antigen Symbol:
    TYRO3
  • Clonality:
    Monoclonal
  • Clone:
    1C10E8 / 9A3F2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.1 mg
  • Environmentally Preferable:
  • Gene ID:
    Q06418
  • Antigen Synonyms:
    TYRO3
  • Storage Buffer:
    Ascitic fluid containing 0.03% sodium azide.
  • Storage Temperature:
    TYRO3 monoclonal antibody can be stored at –20 °C, stable for one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Ni-NTA purified truncated recombinant TYRO3 expressed in E. Coli strain BL21 (DE3).
  • Cat. No.:
    10070-706
  • Supplier no.:
    32-233

Specifications

About this item

Tyrosine-protein kinase (TYRO3) belongs to the Tyr protein kinase family (AXL/UFO subfamily). The UFO family of receptor tyrosine kinases is comprised of subfamily members Rse(also referred to as Tyro3 or Sky) and UFO (also called Tyro7 or Axl). Two distinct isoforms of Rse, designated Brt and Etk-2, have been described. Brt differs from Rse at its C-terminus, but more importantly lacks the N-terminal 31 amino acid signal peptide sequence present in Rse, which is replaced by a 27 amino acid Brt-specific sequence. It has been suggested that as a result of this alternative splicing event, Brt resides in the cytoplasm, unlike Rse which is expressed on the cell surface. Ekt-2 also lacks an N-terminal signal peptide which is substituted with a 45 amino acid Ekt-2-specific sequence. Protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation.

Western Blot:1:500 - 1:2,000
IF:1:50 - 1:200
ELISA:Propose dilution 1:10,000.
Determining optimal working dilutions by titration test.

Ascitic fluid containing 0.03% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TYRO3
Clonality: Monoclonal
Clone: 1C10E8 / 9A3F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human