Anti-GSC Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
GSC regulates chordin (CHRD). GSC may play a role in spatial programing within discrete embryonic fields or lineage compartments during organogenesis.This gene encodes a member of the bicoid subfamily of the paired (PRD) homeobox family of proteins. The encoded protein acts as a transcription factor and may be autoregulatory. A similar protein in mice plays a role in craniofacial and rib cage development during embryogenesis.
GSC antibody can be used for detection of GSC by ELISA at 1:12500. GSC antibody can be used for detection of GSC by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: GSC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
10101-404.pdf
Datasheet
Specifications
- Size:100 ul
- Clonality:Polyclonal
- Molecular Weight:28 kDa
- Purification:Antibody is purified by peptide affinity chromatography method.
- Form:Liquid
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Host:Rabbit
- Cat. No.:10101-404
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:4 °C
- Antibody Type:Primary
- Antigen Symbol:GSC
- Gene ID:P56915
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human GSC.
- Conjugation:Unconjugated
- Antigen Synonyms:GSC
- Antigen Name:goosecoid homeobox
- ELISA:Yes
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store GSC antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.



