This Protein-A purified antibody has been tested for use western blotting and shows specific reactivity by with ESRP2 transfected cell lysates.. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 77.4 kDa.
Recommended Dilutions: ELISA: 1:100000; Western Blot: 1:1000;
Type: Primary
Antigen: ESRP2
Clonality: Monoclonal
Clone: 22C5.H7.A6
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- RL210-301-C32S
- RL210-301-C32
- Antigen Symbol:
- ESRP2
- ESRP2
- Antigen Name:
- Epithelial Splicing Regulatory Protein 2
- Epithelial Splicing Regulatory Protein 2
- Antigen Synonyms:
- 9530027K23Rik|RBM35B
- 9530027K23Rik|RBM35B
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 22C5.H7.A6
- 22C5.H7.A6
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- 77411
- 77411
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Epitope:
- Full length
- Full length
- Immunogen:
- Anti-Esrp-2 is produced by repeated immunizations of full length recombinant mouse Esrp-2 fusion Protein.
- Anti-Esrp-2 is produced by repeated immunizations of full length recombinant mouse Esrp-2 fusion Protein.
- Format:
- Antibody fraction
- Antibody fraction
- Form:
- Liquid (sterile filtered)
- Liquid (sterile filtered)
- Purification:
- Protein A purified
- Protein A purified
- Concentration:
- 1.0 mg/ml by UV absorbance at 280 nm
- 1.0 mg/ml by UV absorbance at 280 nm
- Cross Adsorption:
- No
- No
- Storage Buffer:
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 0.01% (w/v) Sodium Azide,
- Storage Temperature:
- –20 °C
- –20 °C
- Shipping Temperature:
- Dry Ice
- Dry Ice