Level Monitor, Thermo Scientific
Catalog # 490010-934
Supplier: Avantor
Specifications
- Description:Level monitor, LN2, for use with Locator 4 Plus, 230 V
- Environmentally Preferable:
- Cat. No.:490010-934
- Supplier no.:CN509X7
Specifications
About this item
Safeguard irreplaceable biological specimens with the most advanced liquid nitrogen level monitor available. For usage with the Locator 4 Plus cryogenic storage vessel.
- Remotable alarm notifies you of low liquid nitrogen levels even at times when no one is in the building
- LED level indicator panel continuously displays liquid nitrogen volume in 1/8" increments
- Audible and visual alarms provide early warning of low level conditions
- Indicator light flashes when approximately 2 of liquid nitrogen remains in the vessels
- An audible alarm sounds in univision with flashing low-level indicator light when level drops to about 1
- Unlike traditional monitoring devices, this ultrasonic device eliminates contact with liquid nitrogen reducing liquid nitrogen evaporation
3-year warranty