SP Bel-Art Safety Labeled Wash Bottles, Bel-Art Products, a part of SP
Safety labeled wash bottles ensure leakproof dispensing of your most common solvents.
- Leakproof
- Easy-to-read print
- Polypropylene closure construction
- LDPE solvent resistant
Instant recognition, information labeling, and added convenience and safety.
Variety Pack consists of one 500 ml (16 oz.) bottle of each of: Water, Acetone, Methanol, Isopropanol, Toluene, and Ethanol.
*Note: Use only with solvents compatible with LDPE.
Polypropylene closure is 53 mm wide with a leakproof design. Easy-to-read, LDPE solvent resistant, printed bottles include: Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) numbers, protective equipment symbols, and target organ information, making your workplace a safer place.