Polypropylene Graduated Cylinders, Dynalon
: Dynalon
This graduated cylinder is molded of chemically resistant, translucent polypropylene.
- Stable, hexagonal bases with non-slip anti-suction feature
- Very easy to read with virtually no meniscus
Graduations are permanently molded on cylinders and will not fade after repeated autoclaving. Meets stringent international quality criteria according to ISO 9000 standards. Tolerances according to DIN 12 680TT, Class B.
- Catalog No:
- 490007-176
- 490007-178
- Form:
- Tall
- Tall