Specifications
- Chemical method:Winkler
- Environmentally Preferable:
- Cat. No.:89231-358
- Range:1 - 10 mg/L O2
- Supplier no.:2272200
- No. of tests:50
Specifications
About this item
For determination of Dissolved Oxygen by Digital Titrator titration, using the Winkler Azide Modification method. Hach Method 8215. Range: 1 - 10 mg/L Set Includes: Alkaline Iodide-Azide Powder Pillows, Manganous Sulfate Powder Pillows, Sodium Thiosulfate Digital Titrator Cartridge (0.200 N), Starch Indicator Solution and Sulfamic Acid Powder Pillows for approximately 50 tests.
- Accurate Results; when and where you need them
- Fast and Flexible
- Versatile
- Economical