Camel Hair Brushes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Camel hair brushes are used for dusting and cleaning embedding center, balances, and sensitive instruments. Camel hair is mounted on wooden handle with metal ferrule.
- Round, camel hair portion has full body and slightly pointed tip. Brush size 3/4" diameter x 11/2" long
- Paint brush style 1, camel hair portion is 1" long x 1" wide
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.