Stopcock Grease, Ace Glass
Catalog # 89062-802
Supplier: Avantor
Small Business Enterprise
Specifications
- Description:Stopcock Grease
- Environmentally Preferable:
- Cat. No.:89062-802
- Supplier no.:8229-10
Specifications
About this item
A halofluorocarbon lubricant especially suited for lubrication when using strong acids, oxygen, ozone, oxidants, halogens, and other corrosive or reactive chemicals
Has good metal-on-metal lubricating properties and thermal stability up to 260°C. Solid up to 177°C vapor pressure at 25°C… less than 10-3 mm Hg. Complete removal is easily accomplished with organic solvents. 28 gram tube.