To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:C1q and tumor necrosis factor related protein 3
- Antigen Symbol:C1QTNF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:Q9BXJ4
- Antigen Synonyms:C1QTNF3
- Storage Buffer:CTRP3 Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:Predicted: 35 kDa Observed: 36 kDa
- Storage Temperature:CTRP3 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:CTRP3 antibody was raised against a 16 amino acid synthetic peptide from near the center of human CTRP3. The immunogen is located within amino acids 90 - 140 of CTRP3.
- Tested Applications:E, WB, IHC, IF
- Purification:CTRP3 Antibody is affinity chromatography purified via peptide column.
- Cat. No.:89415-868
- Supplier no.:3565
CTRP3 Antibody: Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. Multiple isoforms of human CTRP3 have been reported. It has been suggested that CTRP3 may play a role in skeletal development.
Type: Primary
Antigen: C1QTNF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse