To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:cell death-inducing DFFA-like effector a
- Antigen Symbol:CIDEA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:O60543
- Antigen Synonyms:CIDEA
- Storage Buffer:CIDE-A Antibody is supplied in PBS containing 0.02% sodium azide.
- Molecular Weight:23 kDa
- Storage Temperature:CIDE-A antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Shipping Temperature:4 °C
- Immunogen:CIDE-A antibody was raised against a 18 amino acid peptide near the carboxy terminus of human CIDE-A. The immunogen is located within the last 50 amino acids of CIDE-A.
- Tested Applications:E, WB, IHC, IF
- Purification:CIDE-A Antibody is affinity chromatography purified via peptide column.
- Cat. No.:89415-304
- Supplier no.:2085
CIDE-A Antibody: Apoptosis is related to many diseases and induced by a family of cell death receptors. Cell death signals are transduced by DD-, DED-, or CARD-containing molecules and members of the caspase family of proteases. These death signals finally cause the degradation of chromosomal DNA by activated DNase DFF40/CAD, which is chaperoned and inhibited by DFF45/ICAD. DFF45 related proteins CIDE-A and CIDE-B (for cell death-inducing DFF-like effector A and B) were recently identified. CIDE contains a new type of domain termed CIDE-N, which has high homology with the regulatory domains of DFF45/ICAD and DFF40/CAD. Expression of CIDE-A induces DNA fragmentation and activates apoptosis, which is inhibited by DFF45. CIDE-A is expressed in many tissues.
Type: Primary
Antigen: CIDEA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human