Order Entry
United States
ContactUsLinkComponent
BHI (Brain Heart Infusion) Broth, Hardy Diagnostics
BHI (Brain Heart Infusion) Broth, Hardy Diagnostics
Catalog # 89492-718
Supplier:  Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:  
undefined
BHI (Brain Heart Infusion) Broth, Hardy Diagnostics
Catalog # 89492-718
Supplier:  Hardy Diagnostics
Supplier Number:  R10
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Tube
  • Description:
    Broth
  • Size:
    13 x 100 mm
  • Environmentally Preferable:
  • Storage Temperature:
    2-8C
  • Cat. No.:
    89492-718
  • Fill volume:
    0.5 ml
  • Supplier no.:
    R10

Specifications

About this item

BHI (Brain Heart Infusion) Broth, 0.5ml, 13x100mm tube, optically clear, shatter resistant, polycarbonate tube, order by the package of 20, by Hardy Diagnostics.