Order Entry
United States
ContactUsLinkComponent
Water in Oil Reagent and Apparatus Set, Hach
Water in Oil Reagent and Apparatus Set, Hach
Catalog # 89500-700
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
undefined
Water in Oil Reagent and Apparatus Set, Hach
Catalog # 89500-700
Supplier:  Avantor
Product available on GSA Advantage®
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    Calcium hydride
  • Environmentally Preferable:
  • Method:
    Volumetric Displacement/Calcium Hydride
  • Increments:
    Smallest Step, 0.05%, 0.20%
  • Cat. No.:
    89500-700
  • Range:
    0 - 10% and 0.05 - 1.0%
  • Supplier no.:
    2237300
  • No. of tests:
    25

Specifications

About this item

The Model WO-1 test kit provides critical data in oil quality determinations, oil line operation, and diesel engine maintenance

Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accomodate these items.

The kit has proven particularly useful on board ships. Reagent is packaged in unit-dose vials, and parts in contact with the sample are disposable, so clean-up is eliminated.

Ordering information: Set includes one reagent, test apparatus, and instruction sheet.