Starfish Water-Distribution Manifold with Connector
Catalog # 47800-070
Supplier: Heidolph NA, LLC
Specifications
- Description:Water-distribution manifold with connector
- Environmentally Preferable:
- Cat. No.:47800-070
- Supplier no.:15890340
Specifications
About this item
Water-distribution manifold allows coolant from a single source to be evenly distributed to up to five condensers and then recombines the flow to one outlet pipe.
Two manifolds are used in each set-up-one to distribute water to the condenser and one to collect coolant for recirculation or to drain. Connectors feature leak-proof shut-off valves.