To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:beta-Galactosidase
- Antigen Symbol:b-gal
- Clonality:Polyclonal
- Host:Goat
- Isotype:
- Reactivity:
- Size:2 mL
- Environmentally Preferable:
- Cat. No.:IC56030
- Supplier no.:56030
Anti-β-Galactosidase Host: goat Form: liquid(IgG) Conc/Titer: 3 mg/ml Applications: IFA, cell staining (fluorescent microscopy and cell sorting), tissue staining, and blot immunostaining. There is no cross-reactivity to E.coli. FITC Conjugated